Order Entry
Netherlands
ContactUsLinkComponent
XBridge BEH C8 OBD Prep Column, 130Å, 10 µm, 19 mm X 50 mm 1 * 1 items
Catalog # WATE186004007
Supplier:  WATERS
XBridge BEH C8 OBD Prep Column, 130Å, 10 µm, 19 mm X 50 mm 1 * 1 items
Catalog # WATE186004007
Supplier:  WATERS
Frequently Bought Together