Styragel HMW 2, 500 Å, 20 µm, 7.8 mm X 300 mm in DMF, 100 - 10K 1 * 1 items
Catalog # WATEWAT054494
Supplier: WATERS
Styragel HMW 2, 500 Å, 20 µm, 7.8 mm X 300 mm in DMF, 100 - 10K 1 * 1 items
Catalog # WATEWAT054494
Supplier: WATERS
Frequently Bought Together
VWR®, Screw Caps for Screw Neck Vials ND24
Supplier: VWR Collection
Snap caps for snap cap vials, N 18 and N 22
Supplier: MACHEREY-NAGEL
VWR®, Septa GL 45
Supplier: VWR Collection
Total organic carbon (TOC) vials
Supplier: EP SCIENTIFIC
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
Headspace crimp caps with 20 mm septa
Supplier: Agilent
Screw neck vials, N 9
Supplier: MACHEREY-NAGEL
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
Certified screw top vials and closures
Supplier: Agilent
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
CERT TOC LL <10PPB FOR 40ML GLASS VIALS 1 * 80 items
Supplier: PMT