XTerra MS C18 Prep Column, 125Å, 5 µm, 7.8 mm X 100 mm 1 * 1 items
Catalog # WATE186001156
Supplier: WATERS
XTerra MS C18 Prep Column, 125Å, 5 µm, 7.8 mm X 100 mm 1 * 1 items
Catalog # WATE186001156
Supplier: WATERS
Frequently Bought Together
Safety canisters
Supplier: S.C.A.T.
Vial racks
Supplier: MACHEREY-NAGEL
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Accessories for connection system GLS 80
Supplier: DWK Life Sciences
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Screw neck vials, N 9
Supplier: MACHEREY-NAGEL
Headspace crimp caps with 20 mm septa
Supplier: Agilent
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.