XTerra MS C18 VanGuard Cartridge, 125Å, 2.5 µm, 3.9 mm X 5 mm 1 * 3 items
Catalog # WATE186007889
Supplier: WATERS
XTerra MS C18 VanGuard Cartridge, 125Å, 2.5 µm, 3.9 mm X 5 mm 1 * 3 items
Catalog # WATE186007889
Supplier: WATERS
Frequently Bought Together
Certified screw top vials and closures
Supplier: Agilent
Gastight® 1000 series syringes
Supplier: HAMILTON BONADUZ
Microliter™ 800 series syringes
Supplier: HAMILTON BONADUZ
TSKgel® gel filtration columns, SW-type
Supplier: TOSOH Bioscience
VWR® Air Valve
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND24
Supplier: VWR Collection
Certified headspace crimp top vials
Supplier: Agilent
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
Headspace crimp caps with 20 mm septa
Supplier: Agilent
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
Caps for 20 ml glass crimp top headspace vials, level 1 everyday analysis, SureSTART™
Supplier: Thermo Fisher Scientific
Supplies for Agilent Instruments 7700 Series ICP-MS
Supplier: Agilent
Inlet liners
Supplier: Agilent
TSKgel® gel filtration columns, PW-type
Supplier: TOSOH Bioscience