CORTECS C8 Column, 90Å, 1.6 µm, 3 mm X 30 mm 1 * 3 items
Catalog # WATE176003834
Supplier: WATERS
Frequently Bought Together
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
Safety canisters
Supplier: S.C.A.T.
Certified headspace crimp top vials
Supplier: Agilent
Microliter™ 700 series syringes
Supplier: HAMILTON BONADUZ
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
Headspace crimp caps with 20 mm septa
Supplier: Agilent
Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Supplier: Avantor
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR® Air Valve
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND10
Supplier: VWR Collection
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND15
Supplier: VWR Collection
VWR®, Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection