CORTECS C8 Column, 90Å, 2.7 µm, 3 mm X 30 mm 1 * 3 items
Catalog # WATE176003809
Supplier: WATERS
Frequently Bought Together
Digestion tubes, DigiTUBE®
Supplier: PerkinElmer
SafetyWasteCaps
Supplier: S.C.A.T.
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Caps for 4 ml glass screw top vials, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
VWR®, Septa 12 mm
Supplier: VWR Collection
Glass capillaries and graduated micropipettes, disposable
Supplier: CAMAG
Crimp caps for crimp neck vials, N 20
Supplier: MACHEREY-NAGEL
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Gas-clean filters
Supplier: VARIAN
Safety funnels with lid, 'MARCO' V2.0
Supplier: S.C.A.T.
Screw caps for screw neck vials, N 9
Supplier: MACHEREY-NAGEL
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Vial racks
Supplier: MACHEREY-NAGEL
GC capillary column connectors, Swagelok®
Supplier: PerkinElmer
Microvolume syringes 10 µl
Supplier: Trajan Scientific and Medical
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
Caps for 20 ml glass crimp top headspace vials, level 1 everyday analysis, SureSTART™
Supplier: Thermo Fisher Scientific