CORTECS C18 Column, 90Å, 2.7 µm, 2.1 mm X 30 mm 1 * 3 items
Catalog # WATE176003269
Supplier: WATERS
CORTECS C18 Column, 90Å, 2.7 µm, 2.1 mm X 30 mm 1 * 3 items
Catalog # WATE176003269
Supplier: WATERS
Frequently Bought Together
VWR®, Snap Cap Vials
Supplier: VWR Collection
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Supplier: Avantor
Aluminium oxide for preparative chromatography
Supplier: MERCK PRODUCTION CHEMICALS
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Glass screw top vials, 2 ml, level 1 everyday analysis, SureSTART™
Supplier: Thermo Fisher Scientific
Accessories for syringes
Supplier: Trajan Scientific and Medical
Gas sampling bags, Tedlar®
Supplier: Merck
QuEChERS mixes, clean-up, CHROMABOND®
Supplier: MACHEREY-NAGEL
Accessories for SCAT Safety Caps
Supplier: S.C.A.T.
Capillary column ferrules for gas chromatography
Supplier: Agilent
Size exclusion chromatography columns, OHpak SB series
Supplier: Resonac
Capillaries for LC systems
Supplier: Agilent