Ultrahydrogel DP Column, 120 Å, 6 µm, 7.8 mm X 300 mm, 100 - 5K 1 * 1 items
Catalog # WATEWAT011550
Supplier: WATERS
Ultrahydrogel DP Column, 120 Å, 6 µm, 7.8 mm X 300 mm, 100 - 5K 1 * 1 items
Catalog # WATEWAT011550
Supplier: WATERS
Frequently Bought Together
Screw caps for certified screw top vials
Supplier: Agilent
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh ASTM) for column chromatography, Millipore®
Supplier: MERCK PRODUCTION CHEMICALS
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
VWR®, Vial Racks
Supplier: VWR Collection
VWR®, Septa GL 45
Supplier: VWR Collection
Headspace crimp caps with 20 mm septa
Supplier: Agilent
Exhaust filters, V3.0
Supplier: S.C.A.T.
HPLC columns, LiChrospher®
Supplier: Merck
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Certified screw top vials and closures
Supplier: Agilent
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
CERT TOC LL <10PPB FOR 40ML GLASS VIALS 1 * 80 items
Supplier: PMT