Resolve C8 Radial-Pak Column, 90Å, 10 µm, 5 mm X 100 mm 1 * 1 items
Catalog # WATEWAT085672
Supplier: WATERS
Resolve C8 Radial-Pak Column, 90Å, 10 µm, 5 mm X 100 mm 1 * 1 items
Catalog # WATEWAT085672
Supplier: WATERS
Frequently Bought Together
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Headspace Vials ND18
Supplier: VWR Collection
Certified headspace crimp top vials
Supplier: Agilent
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
Certified microvolume inserts for wide opening vials
Supplier: Agilent
Screw caps for screw neck vials, N 24
Supplier: MACHEREY-NAGEL
Crimp neck vials, N 20
Supplier: MACHEREY-NAGEL
VWR®, Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Gastight® 1700 series syringes
Supplier: HAMILTON BONADUZ
Stopcocks, miniature
Supplier: Bohlender
Amino acids analysis standards, AdvanceBio AAA
Supplier: Agilent
Biodiesel calibration standards
Supplier: Restek
Crimp neck vials, N 11
Supplier: MACHEREY-NAGEL
Storage Boxes for Vials
Supplier: MACHEREY-NAGEL