XSelect Peptide HSS T3 C18 Column, 100Å, 5 µm, 2.1 x 100 mm, with Cytochrome C Digestion Standard 1 * 1 items
WATE176004017
: WATERS
XSelect Peptide HSS T3 C18 Column, 100Å, 5 µm, 2.1 x 100 mm, with Cytochrome C Digestion Standard 1 * 1 items
WATE176004017
: WATERS
Frequently Bought Together
Avantor® ACE® C18, HPLC Columns, 3 µm
: Avantor
ISAspher HPLC Columns, Amino
: ISERA
HPLC columns, NUCLEOSIL® C8
: MACHEREY-NAGEL
Pre-packed columns, 50 and 200 ml, SkillPak™
: TOSOH Bioscience
ISAspher HPLC Columns, Phenyl
: ISERA
Capillary columns, Integra-Guard®
: Restek
Claw and claw holder kit
: Restek
1290 valve head capillaries
: Agilent
Hollow cathode lamps, Atomax™
: PerkinElmer
Avantor® Partisil SAX, HPLC Columns
: Avantor
SPME fiber assemblies, Supelco®
: Merck
Transition fittings, BOLA
: Bohlender
L-MARK® Replacement Plunger for CTC Analytics PAL DLW (Dynamic Load and Wash)
: Trajan Scientific and Medical