XSelect Peptide CSH C18 Prep Column, 130Å, 5 µm, 4.6 mm X 250 mm, 1K - 15K 1 * 1 items
Catalog # WATE186007207
Supplier: WATERS
XSelect Peptide CSH C18 Prep Column, 130Å, 5 µm, 4.6 mm X 250 mm, 1K - 15K 1 * 1 items
Catalog # WATE186007207
Supplier: WATERS
Frequently Bought Together
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Supplier: Avantor
Safety canisters
Supplier: S.C.A.T.
Snap caps for snap cap vials, N 18 and N 22
Supplier: MACHEREY-NAGEL
VWR®, Screw Caps for Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Septa GL 45
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection
Screw neck vials, N 9
Supplier: MACHEREY-NAGEL
Headspace crimp caps with 20 mm septa
Supplier: Agilent
Exhaust filters, V3.0
Supplier: S.C.A.T.