ACQUITY UPLC CSH Fluoro-Phenyl Column, 130Å, 1.7 µm, 3 mm X 150 mm 1 * 1 items
Catalog # WATE186005357
Leverancier: WATERS
ACQUITY UPLC CSH Fluoro-Phenyl Column, 130Å, 1.7 µm, 3 mm X 150 mm 1 * 1 items
Catalog # WATE186005357
Leverancier: WATERS
Frequently Bought Together
Avantor® Ultrasphere®, HPLC Columns
Leverancier: Avantor
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Leverancier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND24
Leverancier: VWR Collection
VWR®, Screw Caps for Screw Neck Headspace Vials ND18
Leverancier: VWR Collection
Microlitre™ syringes for manual HPLC injections on Rheodyne and Valco valves
Leverancier: HAMILTON BONADUZ
Avantor® ACE® C8, HPLC Columns, 5 µm
Leverancier: Avantor
Screw neck vials, N 9
Leverancier: MACHEREY-NAGEL
VWR®, Snap Ring Vials ND11
Leverancier: VWR Collection
VWR®, Snap Cap Vials
Leverancier: VWR Collection
Exhaust filters, V3.0
Leverancier: S.C.A.T.
Avantor® ACE® C18, HPLC Columns, 5 µm
Leverancier: Avantor
Headspace screw neck vials, N 18
Leverancier: MACHEREY-NAGEL
VWR®, Crimp Neck Vials ND11
Leverancier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Leverancier: VWR Collection
Avantor® ACE® C18-AR, HPLC Columns, 3 µm
Leverancier: Avantor
Tubing for peristaltic pumps
Leverancier: PerkinElmer
SPME ASSY 65UM PDMS/DVB NIT 23GA 1 * 3 items
Leverancier: Merck
Empty solid phase extraction columns, CHROMABOND®
Leverancier: MACHEREY-NAGEL
FlashPure EcoFlex Silica Cartridges
Leverancier: BUCHI