Order Entry
Netherlands
ContactUsLinkComponent
CORTECS C8 Column, 90Å, 2.7 µm, 4.6 mm X 150 mm 1 * 1 items
Catalog # WATE186008372
Supplier:  WATERS
CORTECS C8 Column, 90Å, 2.7 µm, 4.6 mm X 150 mm 1 * 1 items
Catalog # WATE186008372
Supplier:  WATERS
Frequently Bought Together