XSelect CSH Phenyl-Hexyl Column, 130Å, 5 µm, 4.6 mm X 50 mm 1 * 1 items
Catalog # WATE186005397
Supplier: WATERS
XSelect CSH Phenyl-Hexyl Column, 130Å, 5 µm, 4.6 mm X 50 mm 1 * 1 items
Catalog # WATE186005397
Supplier: WATERS
Frequently Bought Together
Tubing connectors, b.safe
Supplier: Bohlender
Screw caps, 9 mm, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
ISAspher HPLC Columns, C18 AQ
Supplier: ISERA
Lids for flat bottom chambers
Supplier: CAMAG
Reversed phase HPLC columns, Synergi™ Max-RP
Supplier: PHENOMENEX
Stoppers, 20 mm, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
Safety funnels with lid, 'MARCO' V2.0, electrically conductive
Supplier: S.C.A.T.
Immobilised polysaccharide columns
Supplier: CHIRAL TECHNOLOGIES
Supelco® Ascentis® Express HILIC HPLC and UHPLC Columns
Supplier: Merck
SunFire C18 Column, 100Å, 5µm, 4.6mm x 250mm 1 * 1 items
Supplier: WATERS
Reversed phase HPLC columns, PRP-1
Supplier: HAMILTON BONADUZ
TSKgel® gel filtration columns, PW-type
Supplier: TOSOH Bioscience