XSelect CSH Phenyl-Hexyl Column, 130Å, 5 µm, 2.1 mm X 150 mm 1 * 1 items
Catalog # WATE186005386
Supplier: WATERS
XSelect CSH Phenyl-Hexyl Column, 130Å, 5 µm, 2.1 mm X 150 mm 1 * 1 items
Catalog # WATE186005386
Supplier: WATERS
Frequently Bought Together
VWR®, Screw Caps for Screw Neck Vials ND8
Supplier: VWR Collection
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Avantor® ACE® C8, HPLC Columns, 5 µm
Supplier: Avantor
Glass crimp top headspace vials, 20 ml, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
Microliter™ 700 series syringes
Supplier: HAMILTON BONADUZ
Glass capillaries and graduated micropipettes, disposable
Supplier: CAMAG
Silica gel 60, irregular 40 - 63 µm for flash chromatography
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Screw caps for screw neck vials, N 9
Supplier: MACHEREY-NAGEL
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
Funnel with ball and lid, ARNOLD, V2.0
Supplier: S.C.A.T.
Injection port liners for Agilent Technologies instruments
Supplier: Thermo Fisher Scientific
Canisters, S55, b.safe
Supplier: Bohlender
LC vials and closures
Supplier: Agilent
Supelco® Ascentis® Express HILIC HPLC and UHPLC Columns
Supplier: Merck
Accessories for gas filters Super Clean®
Supplier: Restek
Accessories for chromsep replacement cartridge systems, Pursuit™
Supplier: VARIAN