CORTECS C18+ VanGuard Pre-column, 90Å, 1.6 µm, 2.1 mm X 5 mm 1 * 3 items
Catalog # WATE186007125
Supplier: WATERS
CORTECS C18+ VanGuard Pre-column, 90Å, 1.6 µm, 2.1 mm X 5 mm 1 * 3 items
Catalog # WATE186007125
Supplier: WATERS
Frequently Bought Together
Flash chomatography cartridges, empty colums, CHROMABOND® Flash DL and connecting kits
Supplier: MACHEREY-NAGEL
Crimp neck vials, N 11
Supplier: MACHEREY-NAGEL
SCREW CAP PHENOLIC 15MM BUTYL/PTFE 1 * 100 items
Supplier: DWK Life Sciences
HPLC columns, NUCLEODUR® C18 Gravity
Supplier: MACHEREY-NAGEL
CORTECS C8 Column, 90Å, 2.7 µm, 4.6 mm X 100 mm 1 * 3 items
Supplier: WATERS
ISAspher HPLC Columns, Phenyl
Supplier: ISERA
PTFE/rubber laminated discs
Supplier: Thermo Fisher Scientific
DB-HT SIMDIS 5M 0.53 MM 0.1UM 1 * 1 items
Supplier: Agilent
VWR®, Snap Caps for Snap Ring Vials ND11
Supplier: VWR Collection
Accessories for 7500/7700
Supplier: Agilent
Guard cartridge holder with 3 cartridges 10523 for 10522 1 * 1 items
Supplier: COLLECTION PREFIX HPLC
FlashPure EcoFlex Silica Cartridges
Supplier: BUCHI