XSelect CSH Fluoro-Phenyl Column, 130Å, 3.5 µm, 3 mm X 50 mm 1 * 1 items
Catalog # WATE186005316
Supplier: WATERS
XSelect CSH Fluoro-Phenyl Column, 130Å, 3.5 µm, 3 mm X 50 mm 1 * 1 items
Catalog # WATE186005316
Supplier: WATERS
Frequently Bought Together
ISAspher Reversed Phase HPLC Columns, Amino
Supplier: ISERA
HPLC waste-sets, V2.0
Supplier: S.C.A.T.
Gastight® 1800 series syringes
Supplier: HAMILTON BONADUZ
ISAspher HPLC Columns, Phenyl
Supplier: ISERA
Accessories for 7500/7700
Supplier: Agilent
Starburst plugs for shell vials
Supplier: Restek
Air gas regulators
Supplier: Restek
HPLC columns, Hypersil GOLD™ C18
Supplier: Thermo Fisher Scientific
Symmetry C8 Column, 100Å, 3.5 µm, 3 mm X 100 mm 1 * 1 items
Supplier: WATERS
ISAspher HPLC Columns, PFP
Supplier: ISERA