Order Entry
Netherlands
ContactUsLinkComponent
Symmetry C8 Column, 100Å, 5 µm, 4.6 mm X 100 mm 1 * 1 items
Catalog # WATE186002617
Supplier:  WATERS
Symmetry C8 Column, 100Å, 5 µm, 4.6 mm X 100 mm 1 * 1 items
Catalog # WATE186002617
Supplier:  WATERS
Frequently Bought Together