Viridis CSH Fluoro-Phenyl Column, 130Å, 5 µm, 2.1 mm X 150 mm 1 * 1 items
Catalog # WATE186006543
Supplier: WATERS
Viridis CSH Fluoro-Phenyl Column, 130Å, 5 µm, 2.1 mm X 150 mm 1 * 1 items
Catalog # WATE186006543
Supplier: WATERS
Frequently Bought Together
Avantor® ACE®, Guard Cartridge Holders
Supplier: Avantor
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Supplier: Avantor
HPLC columns, NUCLEOSIL® C18
Supplier: MACHEREY-NAGEL
Canisters, GL 45, b.safe
Supplier: Bohlender
VWR®, Screw Neck Vials ND15
Supplier: VWR Collection
Screw caps for certified screw top vials
Supplier: Agilent
Avantor® ACE® C8, HPLC Columns, 3 µm
Supplier: Avantor
Screw caps with 13 mm septa
Supplier: Agilent
Screw caps for screw neck vials, N 9
Supplier: MACHEREY-NAGEL
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
Avantor® Hichrom, HPLC Columns, RPB
Supplier: Avantor
Glass wool, Supelco®
Supplier: Merck