Viridis CSH Fluoro-Phenyl Column, 130Å, 1.7 µm, 2.1 mm X 75 mm 1 * 1 items
Catalog # WATE186006568
Supplier: WATERS
Viridis CSH Fluoro-Phenyl Column, 130Å, 1.7 µm, 2.1 mm X 75 mm 1 * 1 items
Catalog # WATE186006568
Supplier: WATERS
Frequently Bought Together
VWR®, Screw Caps for Screw Neck Vials ND8
Supplier: VWR Collection
Total organic carbon (TOC) vials
Supplier: EP SCIENTIFIC
Safety canisters
Supplier: S.C.A.T.
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Snap Ring Vials ND11
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR® Standard Micro Syringes
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
Accessories for SCAT Safety Caps
Supplier: S.C.A.T.
Avantor® Alltima, HPLC Columns, C18
Supplier: Avantor
CERT TOC LL <10PPB FOR 40ML GLASS VIALS 1 * 80 items
Supplier: PMT