ACQUITY UPLC M-Class Symmetry C18 Trap Column, 100Å, 5 µm, 180 µm x 20 mm, 2G, V/M 1 * 1 items
Catalog # WATE186007496
Supplier: WATERS
ACQUITY UPLC M-Class Symmetry C18 Trap Column, 100Å, 5 µm, 180 µm x 20 mm, 2G, V/M 1 * 1 items
Catalog # WATE186007496
Supplier: WATERS
Frequently Bought Together
Silica gels for preparative chromatography, standardised and modified
Supplier: MERCK PRODUCTION CHEMICALS
Inlet liners, topaz
Supplier: Restek
ISAspher HPLC Columns, Phenyl
Supplier: ISERA
Syringes, with built-in chaney adapter, Microliter™ and Gastight®
Supplier: HAMILTON BONADUZ
Capillary columns, CP-Sil
Supplier: VARIAN
ISAspher Reversed Phase HPLC Columns, Cyano
Supplier: ISERA
SPME fiber assemblies, Supelco®
Supplier: Merck
Column Acquity In Line Filter Holder and 6 Filters 1 * 1 items
Supplier: WATERS
SPE columns, PCB analysis, CHROMABOND® NAN
Supplier: MACHEREY-NAGEL