ACQUITY UPLC CSH Phenyl-Hexyl Column, 130Å, 1.7 µm, 2.1 mm X 75 mm 1 * 1 items
WATE186005621
: WATERS
ACQUITY UPLC CSH Phenyl-Hexyl Column, 130Å, 1.7 µm, 2.1 mm X 75 mm 1 * 1 items
WATE186005621
: WATERS
Frequently Bought Together
VWR®, Screw Caps for Screw Neck Headspace Vials ND18
: VWR Collection
Certified headspace crimp top vials
: Agilent
VWR®, Storage Boxes for Sample Vials and Bottles, PP
: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
: VWR Collection
Avantor® Ultrasphere®, HPLC Columns
: Avantor
VWR®, Manual Crimper and Decapper
: VWR Collection
VWR®, Snap Cap Vials
: VWR Collection
Silica gel 60, irregular 60 - 200 µm for flash chromatography
: VWR Chemicals
Glass crimp top headspace vials, 20 ml, level 2 high-throughput applications, SureSTART™
: Thermo Fisher Scientific
VWR®, Short Thread Vials ND9
: VWR Collection
TSKgel® gel filtration columns, SW-type
: TOSOH Bioscience
VWR®, Inserts for Short Thread Vials ND9
: VWR Collection
Vials racks
: Agilent
N 20 stopper Bromobutyl, gray null
: MACHEREY-NAGEL