XSelect CSH Fluoro-Phenyl Column, 130Å, 3.5 µm, 1 mm X 50 mm 1 * 1 items
WATE186005304
: WATERS
Frequently Bought Together
VWR®, Manual Crimper and Decapper
: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
: VWR Collection
Glass crimp top headspace vials, 20 ml, level 2 high-throughput applications, SureSTART™
: Thermo Fisher Scientific
VWR®, Screw Neck Headspace Vials ND18
: VWR Collection
VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
: VWR Collection
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
: VWR Collection
Syringeless filters, Mini-UniPrep™, Whatman™
: Whatman products (Cytiva)
Safety canisters
: S.C.A.T.
VWR®, Snap Ring Vials ND11
: VWR Collection
VWR®, Snap Cap Vials
: VWR Collection
Headspace crimp caps with 20 mm septa
: Agilent
Crimp neck vials, N 20
: MACHEREY-NAGEL
Certified screw top vials and closures
: Agilent
VWR®, Crimp Neck Vials ND11
: VWR Collection
N 20 stopper Bromobutyl, gray null
: MACHEREY-NAGEL
Tubing for peristaltic pumps
: PerkinElmer