XBridge BEH C8 Intelligent Speed (IS) Column, 130Å, 3.5 µm, 4.6 mm X 20 mm 1 * 1 items
Catalog # WATE186003183
Supplier: WATERS
XBridge BEH C8 Intelligent Speed (IS) Column, 130Å, 3.5 µm, 4.6 mm X 20 mm 1 * 1 items
Catalog # WATE186003183
Supplier: WATERS
Frequently Bought Together
Silica gels for preparative chromatography, standardised and modified
Supplier: MERCK PRODUCTION CHEMICALS
Avantor® ACE® Excel® C18-PFP, HPLC/UHPLC Columns, Analytical, 2 µm
Supplier: Avantor
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
XSelect CSH C18 Column, 130Å, 5 µm, 3 mm X 100 mm 1 * 1 items
Supplier: WATERS
Avantor® Hichrom HI-DEX B-SE, GC Columns
Supplier: Avantor
Viridis BEH Column, 130Å, 1.7 µm, 2.1 mm X 100 mm 1 * 1 items
Supplier: WATERS
Gas purifiers/filters, Big Trap
Supplier: Trajan Scientific and Medical
Calibration standard, single element, silicon for AAS and ICP-OES, pure grade
Supplier: PerkinElmer
HPLC column tubing
Supplier: Restek
Consumables for Thermo Scientific autosamplers
Supplier: Trajan Scientific and Medical
PE™ C18 Analytical HPLC Columns
Supplier: MICROSOLV