CORTECS Shield RP18 Column, 90Å, 2.7 µm, 2.1 mm X 50 mm 1 * 1 items
Catalog # WATE186008662
Supplier: WATERS
CORTECS Shield RP18 Column, 90Å, 2.7 µm, 2.1 mm X 50 mm 1 * 1 items
Catalog # WATE186008662
Supplier: WATERS
Frequently Bought Together
Avantor® ACE® UltraCore SuperPhenylHexyl, HPLC/UHPLC Columns, 2,5 µm
Supplier: Avantor
Avantor® ACE® C8, HPLC Columns, 5 µm
Supplier: Avantor
Avantor® ACE® C18, HPLC Columns, 3 µm
Supplier: Avantor
Avantor® Partisil Avantor® Partisphere, HPLC Columns
Supplier: Avantor
ISAspher HPLC Columns, Silica, HILIC
Supplier: ISERA
Supplies for fluorescence detector (FLD), G1321A/B 1100/1200 series
Supplier: Agilent
Valves for syringes with valves
Supplier: Trajan Scientific and Medical
CAP 9MM BLU SCRW BND PRSLT PTFE/S 1 * 1.000 items
Supplier: Agilent
POLYSTYRENE STANDARDS KIT, MID-HIGH MW 1 * 1 KIT
Supplier: WATERS
Capillaries for LC systems
Supplier: Agilent
Avantor® ACE® Excel® NH₂, HPLC/UHPLC Columns, Analytical, 1,7 µm
Supplier: Avantor
L-MARK® Replacement Plunger for CTC Analytics PAL-xt
Supplier: Trajan Scientific and Medical
Diamond RSH/RTC syringes
Supplier: Trajan Scientific and Medical
Tubing, flanged, PTFE, BOLA
Supplier: Bohlender
HPLC columns, Acclaim™ Organic Acid
Supplier: Thermo Fisher Scientific