XBridge BEH C8 Column, 130Å, 5 µm, 2.1 mm X 30 mm 1 * 1 items
Catalog # WATE186003187
Supplier: WATERS
XBridge BEH C8 Column, 130Å, 5 µm, 2.1 mm X 30 mm 1 * 1 items
Catalog # WATE186003187
Supplier: WATERS
Frequently Bought Together
VWR®, Vial Racks
Supplier: VWR Collection
VWR®, Septa GL 45
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Snap Ring Vials ND11
Supplier: VWR Collection
Headspace crimp caps with 20 mm septa
Supplier: Agilent
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh ASTM) for column chromatography, Millipore®
Supplier: MERCK PRODUCTION CHEMICALS
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
CERT TOC LL <10PPB FOR 40ML GLASS VIALS 1 * 80 items
Supplier: PMT