ACQUITY UPLC M-Class Peptide CSH C18 Column, 130Å, 1.7 µm, 75 µm X 250 mm 1 * 1 items
Catalog # WATE186007478
Supplier: WATERS
ACQUITY UPLC M-Class Peptide CSH C18 Column, 130Å, 1.7 µm, 75 µm X 250 mm 1 * 1 items
Catalog # WATE186007478
Supplier: WATERS
Frequently Bought Together
Symmetry C8 Column, 100Å, 3.5 µm, 3 mm X 100 mm 1 * 1 items
Supplier: WATERS
XELA UHPLC Columns, C18-AQ-PolE
Supplier: ISERA
SunFire C18 Column, 100Å, 5µm, 4.6mm x 250mm 1 * 1 items
Supplier: WATERS
ISAspher HPLC Columns, SCX
Supplier: ISERA
ISAspher Reversed Phase HPLC Columns, Amino
Supplier: ISERA
Microsyringes for hewlett-packard systems
Supplier: Hospidex
SCREW PLUG 1/8IN PTFE 1 * 2 items
Supplier: Agilent
ISAspher HPLC Columns, C18
Supplier: ISERA
Consumables for Agilent Technologies autosamplers
Supplier: Trajan Scientific and Medical