Order Entry
Netherlands
ContactUsLinkComponent
Atlantis Premier BEH C18 AX Column, 1.7 µm, 2.1 x 50 mm 1 * 1 items
Catalog # WATE186009366
Supplier:  WATERS
Atlantis Premier BEH C18 AX Column, 1.7 µm, 2.1 x 50 mm 1 * 1 items
Catalog # WATE186009366
Supplier:  WATERS
Frequently Bought Together