CORTECS C8 VanGuard Cartridge, 90Å, 2.7 µm, 3.9 mm X 5 mm 1 * 3 items
Catalog # WATE186008422
Supplier: WATERS
CORTECS C8 VanGuard Cartridge, 90Å, 2.7 µm, 3.9 mm X 5 mm 1 * 3 items
Catalog # WATE186008422
Supplier: WATERS
Frequently Bought Together
Tubing connectors, b.safe
Supplier: Bohlender
Avantor® Alltima, HPLC Columns, Amino
Supplier: Avantor
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Avantor® ACE® C18, HPLC Columns, 3 µm
Supplier: Avantor
Microliter™ 700 series syringes
Supplier: HAMILTON BONADUZ
Accessories for 7500/7700
Supplier: Agilent
Accessories for solvent desorption tubes, Supelco®
Supplier: Merck
HyperSep™ Retain™-AX SPE columns
Supplier: Thermo Fisher Scientific
COSMICSIL AGATE 100 C8, 3µm, 150 X 4.6MM 1 * 1 items
Supplier: HICHROM
Spare needles for 5 to 10 ml syringes
Supplier: Trajan Scientific and Medical
Calibration standard, single element, silicon for AAS and ICP-OES, pure grade
Supplier: PerkinElmer
Screw caps for screw neck vials, N 9
Supplier: MACHEREY-NAGEL
Universal waste hubs 'Jan'
Supplier: S.C.A.T.
ISAspher HPLC Columns, Cyano
Supplier: ISERA