Spherisorb C8 OBD Prep Column, 80Å, 10 µm, 10 mm X 250 mm 1 * 1 items
Catalog # WATE186008297
Leverancier: WATERS
Spherisorb C8 OBD Prep Column, 80Å, 10 µm, 10 mm X 250 mm 1 * 1 items
Catalog # WATE186008297
Leverancier: WATERS
Frequently Bought Together
VWR®, Screw Caps for Screw Neck Vials ND24
Leverancier: VWR Collection
Florisil® 60-100 mesh for chromatography
Leverancier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Leverancier: VWR Collection
VWR®, Short Thread Vials ND9
Leverancier: VWR Collection
Air valves for safetycaps generation 2.0
Leverancier: S.C.A.T.
VWR®, Inserts for Short Thread Vials ND9
Leverancier: VWR Collection
VWR®, Manual Crimper and Decapper
Leverancier: VWR Collection
TLC and GLP plates, silica gel 60, unmodified layers, glass backed
Leverancier: Merck
VWR®, Screw Neck Vials ND24
Leverancier: VWR Collection
VWR®, Snap Cap Vials
Leverancier: VWR Collection
Headspace crimp caps with 20 mm septa
Leverancier: Agilent
Exhaust filters, V3.0
Leverancier: S.C.A.T.
VWR®, Crimp Caps for Crimp Neck Vials ND11
Leverancier: VWR Collection
VWR®, Screw Neck Vials ND13
Leverancier: VWR Collection
CERT TOC LL <10PPB FOR 40ML GLASS VIALS 1 * 80 items
Leverancier: PMT