XBridge BEH C8 OBD Prep Column, 130Å, 5 µm, 10 mm X 100 mm 1 * 1 items
Catalog # WATE186008173
Supplier: WATERS
XBridge BEH C8 OBD Prep Column, 130Å, 5 µm, 10 mm X 100 mm 1 * 1 items
Catalog # WATE186008173
Supplier: WATERS
Frequently Bought Together
BND GRN SCR CAP PTFE/WHT SI SEPTA 1 * 100 items
Supplier: Agilent
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Pre-packed columns, 50 and 200 ml, SkillPak™
Supplier: TOSOH Bioscience
ISAspher HPLC Columns, Silica
Supplier: ISERA
Advanced Green 3 Low-Bleed GC Septas
Supplier: Trajan Scientific and Medical
Acrylamide Starter Kit UHPLC EnhClnUp 1 * 1 KIT
Supplier: WATERS
PROTEINWORKS EXPRESS DIGEST START-UP KIT 1 * 1 KIT
Supplier: WATERS
HSPgel Column, 5 µm, 6 mm X 150 mm, 2K - 60K 1 * 1 items
Supplier: WATERS
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
ISAspher HPLC Columns, C4
Supplier: ISERA
LC vials and closures
Supplier: Agilent
Microlab 600 Bubble Free Prime (BFP) syringes
Supplier: HAMILTON BONADUZ
XBridge BEH C8 Column, 130Å, 5 µm, 4.6 mm X 50 mm 1 * 1 items
Supplier: WATERS