XBridge BEH Amide VanGuard Cartridge, 130Å, 5 µm, 2.1 mm X 5 mm 1 * 3 items
Catalog # WATE186007760
Supplier: WATERS
XBridge BEH Amide VanGuard Cartridge, 130Å, 5 µm, 2.1 mm X 5 mm 1 * 3 items
Catalog # WATE186007760
Supplier: WATERS
Frequently Bought Together
Total organic carbon (TOC) vials
Supplier: EP SCIENTIFIC
Guard Cartridge Holder for HPLC Columns
Supplier: Avantor
Screw neck vials, N 8
Supplier: MACHEREY-NAGEL
Glass crimp top headspace vials, 20 ml, level 1 everyday analysis, SureSTART™
Supplier: Thermo Fisher Scientific
Accessories for gas filters Super Clean®
Supplier: Restek
Consumables for Shimadzu autosamplers
Supplier: Trajan Scientific and Medical
LC vials and closures
Supplier: Agilent
Specification certified screw vial and cap kits, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
Syringes for Valco™ VISF-1
Supplier: HAMILTON BONADUZ
PURIFLASH_SILICA HP_30UM_40G_FLASH COLUMN 1 * 20 items
Supplier: Uptima
HPLC columns, NUCLEOSIL® C18
Supplier: MACHEREY-NAGEL
HPLC columns, InfinityLab Poroshell 120 HPH-C18
Supplier: Agilent
Diamond RSH/RTC syringes
Supplier: Trajan Scientific and Medical
Spare Luer lock needles for 10 to 500 µl gas tight syringes
Supplier: Trajan Scientific and Medical