XSelect CSH Fluoro-Phenyl Column, 130Å, 2.5 µm, 2.1 mm X 30 mm 1 * 1 items
Catalog # WATE186006112
Supplier: WATERS
XSelect CSH Fluoro-Phenyl Column, 130Å, 2.5 µm, 2.1 mm X 30 mm 1 * 1 items
Catalog # WATE186006112
Supplier: WATERS
Frequently Bought Together
Certified screw top vials and closures
Supplier: Agilent
Storage Boxes for Vials
Supplier: MACHEREY-NAGEL
Total recovery glass screw top 1,5 ml microvials for <2 ml samples, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Septa GL 45
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Headspace crimp caps with 20 mm septa
Supplier: Agilent
Avantor® Ultrasphere®, HPLC Columns
Supplier: Avantor
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Snap ring vials, N 11
Supplier: MACHEREY-NAGEL
CERT TOC LL <10PPB FOR 40ML GLASS VIALS 1 * 80 items
Supplier: PMT