XSelect CSH Fluoro-Phenyl Prep Guard Cartridge, 130Å, 5 µm, 19 mm X 10 mm 1 * 1 items
Catalog # WATE186005431
Supplier: WATERS
XSelect CSH Fluoro-Phenyl Prep Guard Cartridge, 130Å, 5 µm, 19 mm X 10 mm 1 * 1 items
Catalog # WATE186005431
Supplier: WATERS
Frequently Bought Together
Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, Analytical, 5 µm
Supplier: Avantor
Avantor® ACE® C8, HPLC Columns, 5 µm
Supplier: Avantor
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
FlashPure Select Silica Cartridges
Supplier: BUCHI
b.safe Cap GL 45 PFAS Analytic
Supplier: Bohlender
Tubing, PTFE
Supplier: HAMILTON BONADUZ
Accessories for gas filters Super Clean®
Supplier: Restek
XTerra MS C8 Column, 125Å, 5 µm, 3 mm X 50 mm 1 * 1 items
Supplier: WATERS
Avantor® ACE® C18-AR, HPLC Columns, 3 µm
Supplier: Avantor
Tubing, copper
Supplier: Agilent
Thermo RSH GC autosampler 10 to 50 µl syringe needles
Supplier: Trajan Scientific and Medical
LC vials and closures
Supplier: Agilent
γ-BHC (lindane)
Supplier: Restek
Transition fittings, BOLA
Supplier: Bohlender