XBridge BEH C18 Prep Guard Cartridge, 130Å, 10 µm, 10 mm X 10 mm 1 * 1 items
Catalog # WATE186003889
Supplier: WATERS
XBridge BEH C18 Prep Guard Cartridge, 130Å, 10 µm, 10 mm X 10 mm 1 * 1 items
Catalog # WATE186003889
Supplier: WATERS
Frequently Bought Together
Certified headspace crimp top vials
Supplier: Agilent
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Headspace crimp caps with 20 mm septa
Supplier: Agilent
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
Screw caps for certified screw top vials
Supplier: Agilent
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh ASTM) for column chromatography, Millipore®
Supplier: MERCK PRODUCTION CHEMICALS
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
CERT TOC LL <10PPB FOR 40ML GLASS VIALS 1 * 80 items
Supplier: PMT