XTerra MS C18 Prep Guard Cartridge, 125Å, 10 µm, 7.8 mm X 10 mm 1 * 1 items
Catalog # WATE186001172
Supplier: WATERS
XTerra MS C18 Prep Guard Cartridge, 125Å, 10 µm, 7.8 mm X 10 mm 1 * 1 items
Catalog # WATE186001172
Supplier: WATERS
Frequently Bought Together
Spare needles for autosampler syringes
Supplier: Agilent
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Sb-Cn Rapid Res Hplc Col 4.6 X 75 1 * 1 items
Supplier: Agilent
Drinking water odor standard (2 components)
Supplier: Restek
Rotordicht.,PEEK,3 Rillen,für 0101-0920 1 * 1 items
Supplier: Agilent
ISAspher HPLC Columns, Diol
Supplier: ISERA
Fittings, InfinityLab
Supplier: Agilent
VWR®, Screw Caps for Screw Neck Vials ND18
Supplier: VWR Collection
XBridge BEH C8 Column, 130Å, 5 µm, 4.6 mm X 50 mm 1 * 1 items
Supplier: WATERS