XTerra MS C18 Prep Guard Cartridge, 125Å, 5 µm, 7.8 mm X 10 mm 1 * 1 items
Catalog # WATE186001168
Supplier: WATERS
XTerra MS C18 Prep Guard Cartridge, 125Å, 5 µm, 7.8 mm X 10 mm 1 * 1 items
Catalog # WATE186001168
Supplier: WATERS
Frequently Bought Together
Avantor® ACE® CN, HPLC Columns, 5 µm
Supplier: Avantor
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
Ferrules for gas chromatography
Supplier: Restek
Consumables for Thermo Scientific autosamplers
Supplier: Trajan Scientific and Medical
GC capillary column connectors, Finger Tite
Supplier: Thermo Fisher Scientific
FlashPure Select Silica Cartridges
Supplier: BUCHI
Antistatic explosion proof tubing, b.safe, Bohlender
Supplier: Bohlender
Avantor® ACE® C8, HPLC Columns, 5 µm
Supplier: Avantor
Accessories for 7500/7700
Supplier: Agilent
Acclaim™ Vanquish™ C18 UHPLC Columns
Supplier: Thermo Fisher Scientific
Gastight® 1000 series syringes
Supplier: HAMILTON BONADUZ
Macro cuvettes for spectrophotometers
Supplier: PerkinElmer
HPLC columns for carbohydrate analysis, Hi-Plex
Supplier: VARIAN