BioResolve SEC mAb Column with BioResolve SEC mAb Guard, 200Å, 2.5 µm, 7.8 x 300 mm, with mAb Size Variant Standard 1 * 1 items
Catalog # WATE176004599
Supplier: WATERS
BioResolve SEC mAb Column with BioResolve SEC mAb Guard, 200Å, 2.5 µm, 7.8 x 300 mm, with mAb Size Variant Standard 1 * 1 items
Catalog # WATE176004599
Supplier: WATERS
Frequently Bought Together
Accessories for connection system GLS 80
Supplier: DWK Life Sciences
Certified screw top vials and closures
Supplier: Agilent
TSKgel® gel filtration columns, SW-type
Supplier: TOSOH Bioscience
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Caps for Glass Crimp Top Headspace Vials, Level 3 High Performance Applications, SureSTART™
Supplier: Thermo Fisher Scientific
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
Silica gel 60, irregular 40 - 63 µm for flash chromatography
Supplier: VWR Chemicals
VWR®, Snap Cap Vials
Supplier: VWR Collection
Cellulose microcrystalline for TLC plates
Supplier: Merck
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection