XBridge Protein BEH SEC Column, 200 Å, 2.5 µm, 7.8 mm X 300 mm, and BEH200 SEC Protein Standard Mix 1 * 1 items
WATE176004327
: WATERS
XBridge Protein BEH SEC Column, 200 Å, 2.5 µm, 7.8 mm X 300 mm, and BEH200 SEC Protein Standard Mix 1 * 1 items
WATE176004327
: WATERS
Frequently Bought Together
Gastight® 1000 series syringes
: HAMILTON BONADUZ
Screw caps for screw neck vials, N 9
: MACHEREY-NAGEL
Jumbo syringes
: Trajan Scientific and Medical
Preventive maintenance kit for LC/MS
: Agilent
L-MARK Gold Syringe 50UL Fixed Needle, PE, 51mm Length, 22s Gauge, LC Tip PK6 1 * 6 items
: Trajan Scientific and Medical
Air gas regulators
: Restek
Micro-fluidic platform, SilFlow™
: Trajan Scientific and Medical
XCORE UHPLC Columns, HILIC-Si
: ISERA
VWR® Hichrom C8 HPLC Columns
: Avantor