Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
XSelect Peptide HSS T3 C18 Column, 100Å, 5 µm, 4.6 x 250 mm, with Cytochrome C Digestion Standard 1 * 1 items
WATE176004042
:
WATERS
XSelect Peptide HSS T3 C18 Column, 100Å, 5 µm, 4.6 x 250 mm, with Cytochrome C Digestion Standard 1 * 1 items
WATE176004042
:
WATERS
Frequently Bought Together
Screw caps with 13 mm septa
: Agilent
VWR®, Crimp Neck Vials ND11
: VWR Collection
VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
: VWR Collection
TLC plates, silica gel 60, unmodified and chemically modified layers, aluminium backed
: Merck
Safety canisters
: S.C.A.T.
VWR®, Screw Caps for Screw Neck Vials ND24
: VWR Collection
VWR®, Screw Neck Vials ND13
: VWR Collection
VWR®, Short Thread Vials ND9
: VWR Collection
Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
: Avantor
VWR®, Screw Neck Headspace Vials ND18
: VWR Collection
TLC and GLP plates, silica gel 60, unmodified layers, glass backed
: Merck
VWR®, Screw Caps for Short Thread Vials ND9
: VWR Collection
Exhaust filters, V3.0
: S.C.A.T.
VWR®, Screw Neck Vials ND24
: VWR Collection