ACQUITY UPLC CSH Phenyl-Hexyl Column, 130Å, 1.7 µm, 3 mm X 100 mm 1 * 3 items
Catalog # WATE176002168
Supplier: WATERS
ACQUITY UPLC CSH Phenyl-Hexyl Column, 130Å, 1.7 µm, 3 mm X 100 mm 1 * 3 items
Catalog # WATE176002168
Supplier: WATERS
Frequently Bought Together
VWR®, Screw Caps for Screw Neck Vials ND24
Supplier: VWR Collection
Safety canisters
Supplier: S.C.A.T.
HPLC column accessories
Supplier: Merck
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Accessories for GC/MS
Supplier: Agilent
Snap cap vials, N 18 and N 22
Supplier: MACHEREY-NAGEL
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Accessories for syringes
Supplier: Trajan Scientific and Medical
Syringeless filters, Mini-UniPrep™, Whatman™
Supplier: Whatman products (Cytiva)
Sample head assembly and replacement parts for TurboMatrix HS&Trap
Supplier: PerkinElmer
Snap top vials and closures, 11 mm
Supplier: Agilent
Stopcocks, three-way, Supelco®
Supplier: Merck