ACQUITY UPLC CSH Phenyl-Hexyl Column, 130Å, 1.7 µm, 2.1 mm X 50 mm 1 * 3 items
Catalog # WATE176002163
Supplier: WATERS
ACQUITY UPLC CSH Phenyl-Hexyl Column, 130Å, 1.7 µm, 2.1 mm X 50 mm 1 * 3 items
Catalog # WATE176002163
Supplier: WATERS
Frequently Bought Together
VWR®, Vial Racks
Supplier: VWR Collection
VWR®, Septa GL 45
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Vial racks for headspace vials
Supplier: Restek
Screw caps for certified screw top vials
Supplier: Agilent
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR® Standard Micro Syringes
Supplier: VWR Collection
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
Certified screw top vials and closures
Supplier: Agilent
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
Avantor® Alltima, HPLC Columns, C18
Supplier: Avantor