XTerra MS C8 OBD Prep Column, 125Å, 10 µm, 19 mm X 250 mm 1 * 1 items
Catalog # WATE186002260
Supplier: WATERS
XTerra MS C8 OBD Prep Column, 125Å, 10 µm, 19 mm X 250 mm 1 * 1 items
Catalog # WATE186002260
Supplier: WATERS
Frequently Bought Together
VWR®, Screw Caps for Screw Neck Vials ND8
Supplier: VWR Collection
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
HPLC columns, LiChrospher®
Supplier: Merck
Screw neck vials, N 9
Supplier: MACHEREY-NAGEL
Exhaust filters, V3.0
Supplier: S.C.A.T.
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
TLC plates, silica gel 60, unmodified layers, plastic sheets
Supplier: Merck
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh ASTM) for column chromatography, Millipore®
Supplier: MERCK PRODUCTION CHEMICALS
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
Certified screw top vials and closures
Supplier: Agilent
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
CERT TOC LL <10PPB FOR 40ML GLASS VIALS 1 * 80 items
Supplier: PMT
Tubing for peristaltic pumps
Supplier: PerkinElmer