Sep-Pak Florisil Plus Long Cartridge, 910 mg Sorbent per Cartridge, 50 - 200 µm 1 * 2.000 items
Catalog # WATEWAT036955
Supplier: WATERS
Sep-Pak Florisil Plus Long Cartridge, 910 mg Sorbent per Cartridge, 50 - 200 µm 1 * 2.000 items
Catalog # WATEWAT036955
Supplier: WATERS
Frequently Bought Together
Test strips for peracetic acid, MQuant®, Supelco®
Supplier: Merck
Test strips, peracetic acid, Dosatest
Supplier: VWR Chemicals
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
VWR® Rota®, pH Indicator Paper, 1 to 14
Supplier: VWR Chemicals
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
Aqueous solution containing sulphuric acid (0.001M) and Triton X-100 (0.225% w/v) 1 * 10 L
Supplier: CUSTOM MADE CHEMICALS