TruView pH Control LCMS Certified Amber Glass, 12 x 32 mm, Screw Neck Vial, Max Recovery, with Cap and PTFE/Silicone Septum , 1.5 mL Volume, 1 * 100 items
Catalog # WATE186005664CV
Supplier: WATERS
TruView pH Control LCMS Certified Amber Glass, 12 x 32 mm, Screw Neck Vial, Max Recovery, with Cap and PTFE/Silicone Septum , 1.5 mL Volume, 1 * 100 items
Catalog # WATE186005664CV
Supplier: WATERS
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Sodium hydroxide 2 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
pH electrodes, InLab® ISM
Supplier: Mettler - Toledo
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals