IonHance CX-MS pH Concentrate B pH 8.5 in MS Certified LDPE Container (10X Concentrate) 1 * 1 KIT
Catalog # WATE186009281
Supplier: WATERS
IonHance CX-MS pH Concentrate B pH 8.5 in MS Certified LDPE Container (10X Concentrate) 1 * 1 KIT
Catalog # WATE186009281
Supplier: WATERS
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane ≥98%, SupraSolv® for GC-MS
Supplier: Merck
Cyclohexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Acetonitrile, anhydrous (max. 0.001 % H₂O)
Supplier: Merck
1,4-Dioxane ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetic anhydride for synthesis, Sigma-Aldrich®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
tert-Butyl methyl ether ≥99.0% (by GC), EMPLURA®, Supelco®
Supplier: Merck
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
Pyridine, anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
Ethanol ≥96% (v/v), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Methanol 99.8% for HPLC
Supplier: Thermo Fisher Scientific