2 components: 2000mg/l each of Methylcyclohexane [CAS:108-87-2] 3-Meth 1 * 5 Ampoul
Catalog # CMPLRD0918581.5
Supplier: CUSTOM MADE CHEMICALS LAB
2 components: 2000mg/l each of Methylcyclohexane [CAS:108-87-2] 3-Meth 1 * 5 Ampoul
Catalog # CMPLRD0918581.5
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Test strips, total hardness, MQuant®, Supelco®
Supplier: Merck
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck
Clean-Lab, Q Path® for paraffin removal
Supplier: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test kits, TOC, Spectroquant®, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
Cuvettes and tubes for Picco, Multy, Nova and Prove
Supplier: Merck
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Schiff reagent for histology, Sigma-Aldrich®
Supplier: Merck
Test strips, sulphite, MQuant®, Supelco®
Supplier: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Test kits, photometric, AccuVac®
Supplier: Hach
Test strips, Calcium, QUANTOFIX®
Supplier: MACHEREY-NAGEL