CYCLOPROPANECARBOXYLIC ACID HYDRAZIDE 1 * 5 g
APOSOR54582-5G
Frequently Bought Together
Water, GPR RECTAPUR®
: VWR Chemicals
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Ethanol denatured 96% ≥96%, GPR RECTAPUR®
: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
: VWR Chemicals