Order Entry
Netherlands
Orders LinkContactUsLinkComponent
2-Bromo-3-hydroxypyridine ≥95%

2-Bromo-3-hydroxypyridine ≥95%

Bottle, 25 g
Catalog # APOSOR62149-25G
Supplier:   Apollo Scientific
2-Bromo-3-hydroxypyridine ≥95%
Catalog # APOSOR62149-25G
Supplier:  Apollo Scientific
CAS Number:  6602-32-0
2-Bromo-3-hydroxypyridine ≥95%

Bottle, 25 g

CAS Number:  6602-32-0
Price
21,80 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2-Bromo-3-pyridinol
  • CAS Number:6602-32-0
  • Formula:C₅H₄BrNO
  • MDL Number:MFCD00006220
  • Molecular Weight:174 g/mol
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications