3-(4-BROMOPHENOXY)-6-METHYLPYRIDAZINE 1 * 250 mg
Catalog # APOSOR23343-250MG
Supplier: Apollo Scientific
3-(4-BROMOPHENOXY)-6-METHYLPYRIDAZINE 1 * 250 mg
Catalog # APOSOR23343-250MG
Supplier: Apollo Scientific
Frequently Bought Together
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
Ethyl acetate ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Pyridine, EMPLURA®, Supelco®
Supplier: Merck
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
N-HEPTANE PPA PH-Q UVST 100 LF 1 * 100 L
Supplier: Honeywell Chemicals
Tetrabutylammonium dihydrogen phosphate 0.5 M in HPLC grade water for HPLC
Supplier: Thermo Fisher Scientific
Toluene, extra dry over Molecular Sieve 99.85%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetone, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.9% for HPLC
Supplier: Thermo Fisher Scientific